Skip to Content
Merck
Tetraamminepalladium(II) chloride monohydrate 98%

Tetraamminepalladium(II) chloride monohydrate

Tetraamminepalladium(II) chloride monohydrate 98%
Linear Formula:
Pd(NH3)4Cl2·H2O
CAS Number:
Molecular Weight:
263.46
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing