Skip to Content
Merck
TAGATOSE pharmaceutical secondary standard, certified reference material

Tagatose

TAGATOSE pharmaceutical secondary standard, certified reference material
Synonym(s):
D-(−)-Tagatose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade