Skip to Content
Merck
Stachyose hydrate ≥98% (HPLC)

Stachyose hydrate

Stachyose hydrate ≥98% (HPLC)
Synonym(s):
α-D-Gal-(1→6)-α-D-Gal-(1→6)-α-D-Glc-(1→2)-β-D-Fru, β-D-Fructofuranosyl-O-α-D-galactopyranosyl-(1→6)-O-α-D-galactopyranosyl-(1→6)-α-D-glucopyranoside, Lupeose
Empirical Formula (Hill Notation):
C24H42O21 · xH2O
CAS Number:
Molecular Weight:
666.58 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing