![Spiromesifen PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/653/812/e3c4b4a3-fdd1-4e0e-adb3-30d9adfa0fc7/640/e3c4b4a3-fdd1-4e0e-adb3-30d9adfa0fc7.png)
Spiromesifen
![Spiromesifen PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/653/812/e3c4b4a3-fdd1-4e0e-adb3-30d9adfa0fc7/640/e3c4b4a3-fdd1-4e0e-adb3-30d9adfa0fc7.png)
Synonym(s):
3-Mesityl-2-oxo-1-oxaspiro[4.4]non-3-en-4-yl) 3,3-dimethylbutyrate
Empirical Formula (Hill Notation):
C23H30O4
CAS Number:
Molecular Weight:
370.48
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2