
Sphingosine 1-phosphate

Synonym(s):
(2S,3R,4E)-2-Amino-4-octadecene-1,3-diol 1-phosphate, D-erythro-Sphingosine 1-phosphate
Empirical Formula (Hill Notation):
C18H38NO5P
CAS Number:
Molecular Weight:
379.47
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Optical Isomer
Showing 1-1 of 1