Skip to Content
Merck
Sodium lactate United States Pharmacopeia (USP) Reference Standard

Sodium lactate

Sodium lactate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Sodium L-lactate, (S)-2-Hydroxypropionic acid sodium salt, L-Lactic acid sodium salt, Sarcolactic acid sodium salt
Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing