
Sodium ionophore V

Synonym(s):
4-Octadecanoyloxymethyl-N,N,N′,N′-tetracyclohexyl-1,2-phenylenedioxydiacetamide, ETH 4120
Empirical Formula (Hill Notation):
C53H88N2O6
CAS Number:
Molecular Weight:
849.28
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing