Skip to Content
Merck
Sodium dichloroisocyanurate dihydrate 98.0% (AT)

Sodium dichloroisocyanurate dihydrate

Sodium dichloroisocyanurate dihydrate 98.0% (AT)
Synonym(s):
Dichloroisocyanuric acid sodium salt dihydrate
Empirical Formula (Hill Notation):
C3Cl2N3NaO3 · 2H2O
CAS Number:
Molecular Weight:
255.98
EC Number:

Products

Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing