Skip to Content
Merck
Sodium diatrizoate hydrate ≥98.0%

Sodium diatrizoate hydrate

Sodium diatrizoate hydrate ≥98.0%
Synonym(s):
3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid, sodium salt, 3,5-Diacetamido-2,4,6-triiodobenzoic acid sodium salt, Diatrizoic acid sodium salt hydrate
Empirical Formula (Hill Notation):
C11H8I3N2NaO4 · xH2O
CAS Number:
Molecular Weight:
635.90 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing