Sodium chloroacetate
Synonym(s):
Chloroacetic acid sodium salt, Sodium chloroacetate
Linear Formula:
ClCH2COONa
CAS Number:
Molecular Weight:
116.48
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing