Skip to Content
Merck
Sodium carbonate NIST® SRM® 2192a, pD Standard

Sodium carbonate

Sodium carbonate NIST® SRM® 2192a, pD Standard
Synonym(s):
Calcined soda, Carbonic acid disodium salt, Soda ash
Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Agency
Format
Quality Segment
Special Grade
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing