Skip to Content
Merck
Sodium bicarbonate NIST® SRM® 2191a, pD Standard

Sodium bicarbonate

Sodium bicarbonate NIST® SRM® 2191a, pD Standard
Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Manufacturer
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Reference Material Type
Special Grade
Technique