Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SMCC Conjugation Kit for Iron Oxide (II, III) Nanoparticles
Products
Product Category
kits (4)
Brand
Sigma-Aldrich (4)
Physical Form
Functional Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
OCNIAK15
SMCC Conjugation Kit for Iron Oxide (II, III) Nanoparticles,
15 nm, amine functionalized, 5 mg/mL
Expand
View Pricing
OCNIAK20
SMCC Conjugation Kit for Iron Oxide (II, III) Nanoparticles,
20 nm, amine functionalized, 5 mg/mL
Expand
View Pricing
OCNIAK30
SMCC Conjugation Kit for Iron Oxide (II, III) Nanoparticles,
30 nm, amine functionalized, 5 mg/mL
Expand
View Pricing
OCNIAK25
SMCC Conjugation Kit for Iron Oxide (II, III) Nanoparticles,
25 nm, amine functionalized, 5 mg/mL
Expand
View Pricing