Simvastatin
Synonym(s):
Simvastatin
Empirical Formula (Hill Notation):
C25H38O5
CAS Number:
Molecular Weight:
418.57
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing