![Shogaol ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/378/737/6e6bd2f9-0152-45e1-8006-344ca92937be/640/6e6bd2f9-0152-45e1-8006-344ca92937be.png)
Shogaol
![Shogaol ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/378/737/6e6bd2f9-0152-45e1-8006-344ca92937be/640/6e6bd2f9-0152-45e1-8006-344ca92937be.png)
Synonym(s):
[6]-Shogaol, 1-(4-Hydroxy-3-methoxyphenyl)-4-decen-3-one
Empirical Formula (Hill Notation):
C17H24O3
CAS Number:
Molecular Weight:
276.37
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing