Skip to Content
Merck
(S)-(+)-Ibuprofen ReagentPlus®, 99%

(S)-(+)-Ibuprofen

(S)-(+)-Ibuprofen ReagentPlus®, 99%
Synonym(s):
(S)-(+)-2-(4-Isobutylphenyl)propionic acid, (S)-(+)-4-Isobutyl-α-methylphenylacetic acid
Linear Formula:
(CH3)2CHCH2C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
206.28

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing