
Saxitoxin-15N7 dihydrochloride solution

Empirical Formula (Hill Notation):
C10H1715N7O4 · 2HCl
Molecular Weight:
379.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1