(R,R)-DIPAMP
Synonym(s):
(R,R)-1,2-Bis[(2-methoxyphenyl)(phenylphosphino)]ethane, (R,R)-1,2-Ethanediylbis[(2-methoxyphenyl)phenylphosphine], (R,R)-Ethylenebis[(2-methoxyphenyl)phenylphosphine], [(1R,2R)-(−)-Bis[(2-methoxyphenyl)phenylphosphino]ethane]
Linear Formula:
[CH3OC6H4P(C6H5)CH2-]2
CAS Number:
Molecular Weight:
458.47
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing