![Roxithromycin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/160/850/de96b366-d0e0-4a65-8bfb-b54865ac641b/640/de96b366-d0e0-4a65-8bfb-b54865ac641b.png)
Roxithromycin for system suitability
![Roxithromycin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/160/850/de96b366-d0e0-4a65-8bfb-b54865ac641b/640/de96b366-d0e0-4a65-8bfb-b54865ac641b.png)
Synonym(s):
Roxithromycin, Erythromycin 9-(-O-[2-methoxyethoxy]methyloxime)
Empirical Formula (Hill Notation):
C41H76N2O15
CAS Number:
Molecular Weight:
837.05
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing