Skip to Content
Merck
Rhodium foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%

Rhodium

Rhodium foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Synonym(s):
Rhodium
Empirical Formula (Hill Notation):
Rh
CAS Number:
Molecular Weight:
102.91

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Showing 1-20 of 37
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2