Residual Solvent Class 2 - Methylbutylketone
Synonym(s):
2-Hexanone solution, Butyl methyl ketone
CAS Number:
Products
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing