![Ranolazine Related Compound D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/485/847/7a774a07-b508-4466-8ce6-0e7423295413/640/7a774a07-b508-4466-8ce6-0e7423295413.jpg)
Ranolazine Related Compound D
![Ranolazine Related Compound D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/485/847/7a774a07-b508-4466-8ce6-0e7423295413/640/7a774a07-b508-4466-8ce6-0e7423295413.jpg)
Synonym(s):
2,2′-(Piperazine-1,4-diyl)bis[N-(2,6-dimethylphenyl)acetamide], N1,N4-Bis(2,6-dimethylphenyl)-1,4-piperazinediacetamide
Empirical Formula (Hill Notation):
C24H32N4O2
CAS Number:
Molecular Weight:
408.54
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2