![Raltegravir potassium European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/369/813/1da49db6-f695-489d-a13b-57d9c1d156e7/640/1da49db6-f695-489d-a13b-57d9c1d156e7.png)
Raltegravir potassium
![Raltegravir potassium European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/369/813/1da49db6-f695-489d-a13b-57d9c1d156e7/640/1da49db6-f695-489d-a13b-57d9c1d156e7.png)
Synonym(s):
Raltegravir potassium
Empirical Formula (Hill Notation):
C20H20FKN6O5
CAS Number:
Molecular Weight:
482.51
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3