(R)-(−)-2-Chloromandelic acid
Linear Formula:
ClC6H4CH(OH)CO2H
CAS Number:
Molecular Weight:
186.59
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing