Skip to Content
Merck
(R)-(−)-1-Aminoindane 97%

(R)-(−)-1-Aminoindane

(R)-(−)-1-Aminoindane 97%
Synonym(s):
(R)-(−)-1-Indanamine
Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing