Skip to Content
Merck
(R)-(−)-1-(9-Anthryl)-2,2,2-trifluoroethanol ≥98%

(R)-(−)-1-(9-Anthryl)-2,2,2-trifluoroethanol

(R)-(−)-1-(9-Anthryl)-2,2,2-trifluoroethanol ≥98%
Synonym(s):
(R)-(−)-α-(Trifluoromethyl)anthracene-9-methanol, (R)-(−)-2,2,2-Trifluoro-1-(9-anthryl)ethanol
Empirical Formula (Hill Notation):
C16H11F3O
CAS Number:
Molecular Weight:
276.25
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing