Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quinine monohydrochloride dihydrate
Synonym(s):
Quinine monohydrochloride dihydrate, Quinine hydrochloride dihydrate
Empirical Formula (Hill Notation):
C
20
H
24
N
2
O
2
· HCl · 2H
2
O
CAS Number:
6119-47-7
Molecular Weight:
396.91
EC Number:
205-001-1
Products
Product Category
aroma chemicals (1)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (1)
Halal (1)
Kosher (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
W297607
Quinine monohydrochloride dihydrate,
≥95%, FG
Expand
View Pricing
145920
Quinine monohydrochloride dihydrate,
90%
Expand
View Pricing
V900715
Quinine monohydrochloride dihydrate,
Vetec
™
, reagent grade, 88%
Expand
View Pricing