Skip to Content
Merck
Quinine hemisulfate salt monohydrate tested according to Ph. Eur.

Quinine hemisulfate salt monohydrate

Quinine hemisulfate salt monohydrate tested according to Ph. Eur.
Synonym(s):
Quinine sulfate (2:1) (salt) dihydrate
Empirical Formula (Hill Notation):
C20H24N2O2 · 0.5H2O4S · H2O
CAS Number:
Molecular Weight:
391.47

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
Product Line
Purity
Activity Spectrum
Agency
Quality Segment
Markush Class
Markush Group
Mode of Action
Special Grade