Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Prostaglandin E
1
Synonym(s):
(11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE
1
, Prostaglandin E
1
Empirical Formula (Hill Notation):
C
20
H
34
O
5
CAS Number:
745-65-3
Molecular Weight:
354.48
EC Number:
212-017-2
Products
Product Category
bioactive small molecules (2)
hormones (2)
Brand
Millipore (1)
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Research Area
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P5515
Prostaglandin E
1
,
≥98% (HPLC), powder, vasodilator
Expand
View Pricing
P7527
Prostaglandin E
1
,
γ-irradiated, powder, BioXtra, suitable for cell culture
Expand
View Pricing
538903
Prostaglandin E
1
,
Activates adenylate cyclase activity via a G-protein coupled receptor.
Expand
View Pricing
P8908
Prostaglandin E
1
,
synthetic, powder, BioReagent, suitable for cell culture
Expand
View Pricing