Propafenone Related Compound A
Synonym(s):
N-{2-Hydroxy-3-[2-(3-phenylpropanoyl)phenoxy]propyl}-N-propylformamide
Empirical Formula (Hill Notation):
C22H27NO4
Molecular Weight:
369.45
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2