Propafenone hydrochloride
Synonym(s):
1-[2-(2-Hydroxy-3-(propylamino)propoxy)phenyl]-3-phenyl-1-propanone hydrochloride
Empirical Formula (Hill Notation):
C21H27NO3 · HCl
CAS Number:
Molecular Weight:
377.90
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3