Skip to Content
Merck
Potassium ferricyanide(III) powder or chunks, <10 μm, 99%

Potassium ferricyanide(III)

Potassium ferricyanide(III) powder or chunks, <10 μm, 99%
Synonym(s):
Potassium hexacyanoferrate(III), Red prussiate
Linear Formula:
K3Fe(CN)6
CAS Number:
EC Number:

Products

Product Category
Brand
Physical Form
Purity
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing