Skip to Content
Merck
Platinum(IV) oxide hydrate crystalline, 99.5%

Platinum(IV) oxide hydrate

Platinum(IV) oxide hydrate crystalline, 99.5%
Synonym(s):
Platinum(IV) oxide hydrate (80% Pt)
Linear Formula:
PtO2 · xH2O
CAS Number:
Molecular Weight:
227.08 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Core