Plastic additive 5
Synonym(s):
Tris(2,4-di-tert-butylphenyl) phosphite
Linear Formula:
[[(CH3)3C]2C6H3O]3P
CAS Number:
Molecular Weight:
646.92
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2