Pirfenidone
Synonym(s):
5-Methyl-1-phenyl-2-(1H)-pyridone
Empirical Formula (Hill Notation):
C12H11NO
CAS Number:
Molecular Weight:
185.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing