![Piperazine Dihydrochloride (Piperazine Dihydrochloride Hydrate) certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/107/423/ee2a9a73-e516-4e07-9bf5-1ab77a757215/640/ee2a9a73-e516-4e07-9bf5-1ab77a757215.jpg)
Piperazine Dihydrochloride (Piperazine Dihydrochloride Hydrate)
![Piperazine Dihydrochloride (Piperazine Dihydrochloride Hydrate) certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/107/423/ee2a9a73-e516-4e07-9bf5-1ab77a757215/640/ee2a9a73-e516-4e07-9bf5-1ab77a757215.jpg)
Synonym(s):
Piperazine dihydrochloride
Empirical Formula (Hill Notation):
C4H10N2 · 2HCl · xH2O
CAS Number:
Molecular Weight:
159.06 (anhydrous basis)
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing