Skip to Content
Merck
vibrant-m

Piperazine dihydrochloride

vibrant-m
Empirical Formula (Hill Notation):
C4H10N2 · 2HCl · xH2O
CAS Number:
Molecular Weight:
159.06 (anhydrous basis)

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1