Skip to Content
Merck
Pheophorbide-a ≥90% (HPLC)

Pheophorbide-a

Pheophorbide-a ≥90% (HPLC)
Synonym(s):
pheophorbide a
Empirical Formula (Hill Notation):
C35H36N4O5
CAS Number:
Molecular Weight:
592.68

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing