Skip to Content
Merck
Phenylacetaldehyde ≥95%, FCC, FG

Phenylacetaldehyde

Phenylacetaldehyde ≥95%, FCC, FG
Synonym(s):
α-Tolyaldehyde
Linear Formula:
C6H5CH2CHO
CAS Number:
Molecular Weight:
120.15
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing