Skip to Content
Merck
Phenol-d6 99 atom % D

Phenol-d6

Phenol-d6 99 atom % D
Synonym(s):
Hexadeuterophenol
Linear Formula:
C6D5OD
CAS Number:
Molecular Weight:
100.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing