![Perfluorotetradecanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/327/847/e5ce22c5-483f-4552-a9d9-9e343478880c/640/e5ce22c5-483f-4552-a9d9-9e343478880c.png)
Perfluorotetradecanoic acid
![Perfluorotetradecanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/327/847/e5ce22c5-483f-4552-a9d9-9e343478880c/640/e5ce22c5-483f-4552-a9d9-9e343478880c.png)
Linear Formula:
CF3(CF2)12CO2H
CAS Number:
Molecular Weight:
714.11
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2