Skip to Content
Merck
Perfluoro-3,6-dioxaheptanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Perfluoro-3,6-dioxaheptanoic acid

Perfluoro-3,6-dioxaheptanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2,2-Difluoro-2-[1,1,2,2-tetrafluoro-2-(trifluoromethoxy)ethoxy]acetic acid, Nonafluoro-3,6-dioxaheptanoic acid
Empirical Formula (Hill Notation):
C5HF9O4
CAS Number:
Molecular Weight:
296.04

Products

Brand
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade