Skip to Content
Merck
PCB No 31 analytical standard

PCB No 31

PCB No 31 analytical standard
Synonym(s):
2,4′,5-Trichlorobiphenyl
Empirical Formula (Hill Notation):
C12H7Cl3
CAS Number:
Molecular Weight:
257.54

Products

Brand
Formula Weight
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing