![Parbendazole VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/664/458/73667e5c-03d9-4a86-9aed-6cd41ba2f430/640/73667e5c-03d9-4a86-9aed-6cd41ba2f430.png)
Parbendazole
![Parbendazole VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/664/458/73667e5c-03d9-4a86-9aed-6cd41ba2f430/640/73667e5c-03d9-4a86-9aed-6cd41ba2f430.png)
Synonym(s):
Methyl (5-butyl-1H-benzoimidazol-2-yl)carbamate, Methyl 5(6)-butyl-2-benzimidazolecarbamate
Empirical Formula (Hill Notation):
C13H17N3O2
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing