
Parathyroid hormone human

Synonym(s):
PTH, Parathormone
Empirical Formula (Hill Notation):
C408H674N126O126S2
CAS Number:
Molecular Weight:
9424.62
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Sterility
Showing 1-1 of 1