![Pararosaniline (chloride) (C.I. 42500)](/deepweb/assets/sigmaaldrich/product/images/334/195/803cf325-a71e-48a0-80e0-93fc970d02b4/640/803cf325-a71e-48a0-80e0-93fc970d02b4.jpg)
Pararosaniline (chloride) (C.I. 42500)
![Pararosaniline (chloride) (C.I. 42500)](/deepweb/assets/sigmaaldrich/product/images/334/195/803cf325-a71e-48a0-80e0-93fc970d02b4/640/803cf325-a71e-48a0-80e0-93fc970d02b4.jpg)
Empirical Formula (Hill Notation):
C19H18ClN3
CAS Number:
Molecular Weight:
323.82
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing