![Paraquat dichloride hydrate PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/199/452/7b43277b-d436-4986-bac2-b558784322b7/640/7b43277b-d436-4986-bac2-b558784322b7.png)
Paraquat dichloride hydrate
![Paraquat dichloride hydrate PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/199/452/7b43277b-d436-4986-bac2-b558784322b7/640/7b43277b-d436-4986-bac2-b558784322b7.png)
Synonym(s):
1,1′-Dimethyl-4,4′-bipyridinium dichloride hydrate, Gramoxone, Methyl viologen dichloride, Paraquat dichloride
Empirical Formula (Hill Notation):
C12H14Cl2N2 · xH2O
CAS Number:
Molecular Weight:
257.16 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing