Oxibendazole
Synonym(s):
Methyl (5-propoxy-1H-benzimidazol-2-yl)carbamate
Empirical Formula (Hill Notation):
C12H15N3O3
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing