
Oxeladin hydrogen citrate

Synonym(s):
Oxeladin citrate salt
Empirical Formula (Hill Notation):
C20H33NO3 · C6H8O7
CAS Number:
Molecular Weight:
527.60
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing