Oxacillin sodium
Synonym(s):
Oxacillin sodium salt monohydrate
Linear Formula:
C19H18N3O5SNa · H2O
CAS Number:
Molecular Weight:
441.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2